Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017401.2 | NP_059097.2 | 5 | 952 | ||
GenBank mRNA | AF176098.1 | - | 5 | 725 | ||
AJ251804.2 | - | 5 | 696 | |||
AK137747.1 | - | 3 | 2017 | |||
BC056213.1 | - | 5 | 744 | 30024844 | ||
BC056996.1 | - | 2 | 2599 |