Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016881.2 | NP_058577.1 | 6 | 515 | ||
GenBank mRNA | AF043514.1 | - | 6 | 515 | ||
AK002858.1 | - | 6 | 457 | |||
AK003334.1 | - | 6 | 473 | |||
AK005074.1 | - | 6 | 497 | |||
AK012683.1 | - | 6 | 499 | |||
AK134850.1 | - | 467 | ||||
AK145402.1 | - | 6 | 467 | |||
AK149116.1 | - | 6 | 477 | |||
BC037101.1 | - | 6 | 492 | |||
BC046325.1 | - | 6 | 463 | 6395635 | ||
BC094448.1 | - | 6 | 481 | 4238664 |