Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016860.1 | NP_058556.1 | 3 | 188 | ||
GenBank mRNA | AB010297.1 | - | 3 | 188 | ||
AK010632.1 | - | 3 | 185 | |||
AK019518.1 | - | 3 | 192 | |||
AK088062.1 | - | 3 | 195 | |||
AK162248.1 | - | 3 | 202 | |||
AK167450.1 | - | 3 | 195 | |||
AK168889.1 | - | 3 | 192 | |||
AK206467.1 | - | 2 | 77 | |||
BC007131.1 | - | 3 | 203 | 3599656 |