Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016860.1 | NP_058556.1 | 7 | 728 | ||
GenBank mRNA | AB010297.1 | - | 7 | 728 | ||
AK010632.1 | - | 7 | 725 | |||
AK019518.1 | - | 7 | 732 | |||
AK088062.1 | - | 7 | 735 | |||
AK162248.1 | - | 7 | 743 | |||
AK167450.1 | - | 7 | 735 | |||
AK168889.1 | - | 7 | 732 | |||
AK182156.1 | - | 3 | 268 | |||
BC007131.1 | - | 7 | 743 | 3599656 |