Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016861.4 | NP_058557.2 | 4 | 622 | ||
GenBank mRNA | AF053367.2 | - | 4 | 538 | ||
AK076285.1 | - | 521 | ||||
AK088089.1 | - | 4 | 521 | |||
AK146225.1 | - | 4 | 532 | |||
AK146399.1 | - | 4 | 534 | |||
AK158173.1 | - | 4 | 521 | |||
AK189667.1 | - | 2 | 153 | |||
BC004809.1 | - | 4 | 513 | 3588132 |