Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019397.3 | NP_062270.1 | 11 | 1660 | ||
GenBank mRNA | AJ245672.1 | - | 11 | 1662 | ||
BC117702.2 | - | 11 | 1646 | 40062530 |