Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017478.3 | NP_059506.1 | 879 | |||
GenBank mRNA | AF205065.1 | - | 782 | |||
AJ251067.1 | - | 851 | ||||
AK079599.1 | - | 11 | 904 | |||
AK088492.1 | - | 863 | ||||
AK147943.1 | - | 825 | ||||
AK163668.1 | - | 10 | 850 | |||
BC049178.1 | - | 845 | 6309945 | |||
BC145898.1 | - | 822 | 40131201 | |||
BC145900.1 | - | 822 | 40131203 |