Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017480.2 | NP_059508.2 | 4 | 575 | ||
GenBank mRNA | AB023132.1 | - | 4 | 559 | ||
AF216748.1 | - | 4 | 530 | |||
AF257230.1 | - | 4 | 560 | |||
AJ250559.1 | - | 4 | 575 | |||
AK030827.1 | - | 4 | 568 | |||
BC034852.1 | - | 4 | 562 | 1364228 |