Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205241.1 | NP_001192170.1 | 14, 15 | 2451 | ||
NM_017479.3 | NP_059507.2 | 14, 15 | 2575 | |||
GenBank mRNA | AF222800.1 | - | 14, 15 | 2491 | ||
AK045188.1 | - | 14, 15 | 2914 | |||
AK147509.1 | - | 14, 15 | 2452 | |||
BC095974.1 | - | 13, 14 | 2228 |