Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rragc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017475.2 | NP_059503.2 | 2 | 518 | ||
GenBank mRNA | AB017616.1 | - | 2 | 441 | ||
AK150792.1 | - | 2 | 472 | |||
AK159355.1 | - | 2 | 451 | |||
AK159434.1 | - | 2 | 473 | |||
AK166658.1 | - | 2 | 464 | |||
BC005417.1 | - | 2 | 466 | 3495022 | ||
BC071245.1 | - | 2 | 477 |