Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019402.2 | NP_062275.1 | 3, 4 | 546 | ||
GenBank mRNA | AK032172.1 | - | 3, 4 | 542 | ||
AK157143.1 | - | 3, 4 | 535 | |||
BC049869.1 | - | 3, 4 | 531 | |||
BC055866.1 | - | 3, 4 | 535 | 4218469 |