Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019403.3 | NP_062276.1 | 5 | 516 | ||
GenBank mRNA | AK010792.1 | - | 5 | 546 | ||
AK012729.1 | - | 5 | 508 | |||
AK020514.2 | - | 5 | 517 | |||
AK020515.2 | - | 5 | 535 | |||
BC012031.1 | - | 5 | 468 | |||
BC016449.1 | - | 5 | 516 | 4191746 |