Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl6ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019419.2 | NP_062292.1 | 6 | 646 | ||
GenBank mRNA | AF172088.1 | - | 6 | 632 | ||
AF223953.1 | - | 6 | 584 | |||
AK090001.1 | - | 6 | 598 | |||
AK150042.1 | - | 6 | 599 | |||
AK150507.1 | - | 6 | 597 | |||
AK151046.1 | - | 4 | 364 | |||
AK151272.1 | - | 6 | 596 | |||
AK151507.1 | - | 6 | 596 | |||
AK152565.1 | - | 6 | 596 | |||
AK152885.1 | - | 6 | 593 | |||
AK154355.1 | - | 6 | 618 | |||
AK154644.1 | - | 6 | 599 | |||
AK163655.1 | - | 6 | 596 | |||
AK167872.1 | - | 6 | 601 | |||
AK167888.1 | - | 6 | 649 | |||
AK168172.1 | - | 6 | 599 | |||
AK168245.1 | - | 6 | 598 | |||
AK169191.1 | - | 6 | 596 | |||
BC010196.1 | - | 6 | 586 | 3157834 | ||
BC046792.1 | - | 6 | 569 | 5717527 |