Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl6ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019419.2 | NP_062292.1 | 4 | 433 | ||
GenBank mRNA | AF133669.1 | - | 4 | 378 | ||
AF172088.1 | - | 4 | 419 | |||
AF223953.1 | - | 4 | 371 | |||
AK090001.1 | - | 4 | 385 | |||
AK150042.1 | - | 4 | 386 | |||
AK150507.1 | - | 4 | 384 | |||
AK151046.1 | - | 2 | 151 | |||
AK151272.1 | - | 4 | 383 | |||
AK151507.1 | - | 4 | 383 | |||
AK152565.1 | - | 4 | 383 | |||
AK152885.1 | - | 4 | 380 | |||
AK154355.1 | - | 4 | 405 | |||
AK154644.1 | - | 4 | 386 | |||
AK163655.1 | - | 4 | 383 | |||
AK167872.1 | - | 4 | 388 | |||
AK167888.1 | - | 4 | 436 | |||
AK167912.1 | - | 4 | 383 | |||
AK168172.1 | - | 4 | 386 | |||
AK168245.1 | - | 4 | 385 | |||
AK169191.1 | - | 4 | 383 | |||
AK176958.1 | - | 4 | 295 | |||
AK217186.1 | - | 4 | 292 | |||
BC010196.1 | - | 4 | 373 | 3157834 | ||
BC046792.1 | - | 4 | 356 | 5717527 | ||
EF437380.1 | - | 4 | 317 |