Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc23a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018824.2 | NP_061294.2 | 12 | 1535 | ||
GenBank mRNA | AB038145.1 | - | 11 | 1271 | ||
AK122227.1 | - | 8 | 1307 | |||
AY004874.1 | - | 12 | 1489 | |||
BC050823.1 | - | 11 | 1299 | 6332451 |