Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018826.2 | NP_061296.1 | 3 | 1133 | ||
GenBank mRNA | AF165985.1 | - | 3 | 745 | ||
AF230074.1 | - | 3 | 1300 | |||
AJ271054.1 | - | 3 | 526 | |||
AK004747.1 | - | 3 | 946 | |||
BC051959.1 | - | 3 | 926 | 6404946 | ||
BC056994.1 | - | 3 | 1133 | 5716910 |