Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005554.3 | NP_005545.1 | 9 | 2200 | ||
GenBank mRNA | AK299386.1 | - | 10 | 1972 | ||
BC008807.2 | - | 9 | 2039 | 3947610 | ||
BC014152.2 | - | 9 | 2043 | 3639270 | ||
BC069269.1 | - | 9 | 2021 | 5428084 |
Gene Symbol: KRT6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173086.4 | NP_775109.2 | 9 | 2043 | ||
GenBank mRNA | BC110639.1 | - | 9 | 2043 | 4754834 |