Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018755.2 | NP_061225.2 | 9 | 1574 | ||
NM_176073.4 | NP_788262.1 | 8 | 1484 | |||
GenBank mRNA | AF009513.1 | - | 8 | 1329 | ||
AF107835.1 | - | 8 | 1452 | |||
AK032972.1 | - | 8 | 1349 | |||
AK075686.1 | - | 8 | 1415 | |||
AK135096.1 | - | 8 | 1471 | |||
AK137164.1 | - | 8 | 1358 | |||
AK152139.1 | - | 9 | 1452 | |||
AK152145.1 | - | 9 | 1452 | |||
BC010977.1 | - | 8 | 1469 | |||
BC037067.1 | - | 8 | 1331 | 4989536 |