Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm3ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019434.2 | NP_062307.2 | 25 | 5571 | ||
GenBank mRNA | AJ006590.1 | - | 25 | 5604 | ||
AK052382.1 | - | 20 | 3427 | |||
AK122313.1 | - | 25 | 5615 | |||
AK165254.1 | - | 14 | 2276 | |||
BC052452.1 | - | 25 | 5571 | 6409827 | ||
BC067414.1 | - | 24 | 4506 |