Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019436.3 | NP_062309.2 | 2 | 258 | ||
GenBank mRNA | AJ236881.1 | - | 2 | 234 | ||
AK079882.1 | - | 2 | 260 | |||
BC064055.1 | - | 2 | 271 | 576098 |