Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018753.6 | NP_061223.2 | 5 | 804 | ||
GenBank mRNA | AF058797.1 | - | 5 | 803 | ||
AK002632.1 | - | 5 | 823 | |||
AK004872.1 | - | 5 | 805 | |||
AK011389.1 | - | 5 | 804 | |||
AK083367.1 | - | 5 | 794 | |||
AK144061.1 | - | 5 | 867 | |||
AK150414.1 | - | 5 | 817 | |||
AK151294.1 | - | 5 | 817 | |||
AK158932.1 | - | 5 | 707 | |||
AK178774.1 | - | 4 | 364 | |||
AK186637.1 | - | 4 | 365 | |||
AK188420.1 | - | 3 | 225 | |||
AK206680.1 | - | 3 | 336 | |||
AK206731.1 | - | 3 | 225 | |||
AK213523.1 | - | 3 | 225 | |||
AK217062.1 | - | 3 | 347 | |||
AK217335.1 | - | 3 | 225 | |||
AK219160.1 | - | 3 | 336 | |||
BC049070.1 | - | 638 | ||||
BC132121.1 | - | 5 | 658 | 40130398 | ||
BC138512.1 | - | 5 | 658 | 8861532 |