Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018753.6 | NP_061223.2 | 2 | 377 | ||
GenBank mRNA | AF058797.1 | - | 2 | 376 | ||
AK002632.1 | - | 2 | 396 | |||
AK004872.1 | - | 2 | 378 | |||
AK011389.1 | - | 2 | 377 | |||
AK083367.1 | - | 2 | 367 | |||
AK144061.1 | - | 2 | 440 | |||
AK150414.1 | - | 2 | 390 | |||
AK151294.1 | - | 2 | 390 | |||
AK158932.1 | - | 2 | 280 | |||
BC049070.1 | - | 211 | ||||
BC132121.1 | - | 2 | 231 | 40130398 | ||
BC138512.1 | - | 2 | 231 | 8861532 |