Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019443.2 | NP_062316.1 | 3 | 439 | ||
GenBank mRNA | AK007840.1 | - | 3 | 346 | ||
AK131584.1 | - | 638 | ||||
BC024364.1 | - | 3 | 348 | 5031206 | ||
CF383819.1 | - | 2 | 223 | |||
Y07708.1 | - | 3 | 458 |