Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019445.2 | NP_062318.2 | 10 | 4109 | ||
GenBank mRNA | AF218940.1 | - | 11 | 4071 | ||
AK013585.1 | - | 5 | 358 | |||
AK140268.1 | - | 4 | 394 | |||
BC094606.1 | - | 10 | 4109 | 6840339 |