Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Barhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164186.1 | NP_001157658.1 | 4 | 1258 | ||
NM_019446.4 | NP_062319.1 | 3 | 1541 | |||
GenBank mRNA | AB043980.1 | - | 3 | 1556 | ||
AF264026.1 | - | 5 | 1316 | |||
AJ237590.1 | - | 3 | 816 | |||
AK036312.1 | - | 3 | 993 | |||
AK048063.1 | - | 3 | 394 | |||
BC055731.1 | - | 4 | 1259 | 6413504 |