Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgfac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019447.2 | NP_062320.2 | 11 | 1422 | ||
GenBank mRNA | AF099017.1 | - | 11 | 1382 | ||
AF224724.1 | - | 11 | 1382 | |||
AK007987.1 | - | 11 | 1420 | |||
AK075718.1 | - | 11 | 1331 | |||
AK144554.1 | - | 11 | 1393 | |||
AK149522.1 | - | 11 | 1386 | |||
AK165433.1 | - | 9 | 1771 | |||
AK169989.1 | - | 11 | 1418 | |||
BC019376.1 | - | 11 | 1387 | 5134851 |