Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018830.1 | NP_061300.1 | 7 | 815 | ||
GenBank mRNA | AB037111.1 | - | 8 | 1100 | ||
AB037181.1 | - | 7 | 815 | |||
AK034493.1 | - | 7 | 1046 | |||
AK041937.1 | - | 7 | 1004 | |||
AK046540.1 | - | 7 | 1009 | |||
AK047692.1 | - | 7 | 1003 | |||
AK080951.1 | - | 7 | 992 | |||
AK166100.1 | - | 7 | 1063 | |||
BC137898.1 | - | 7 | 798 | 8860917 | ||
BC145016.1 | - | 7 | 801 | 9053564 |