Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018810.2 | NP_061280.2 | 6 | 1110 | ||
GenBank mRNA | AB026986.1 | - | 6 | 1215 | ||
AF192785.1 | - | 6 | 1135 | |||
AK004225.1 | - | 1 | 16 | |||
AK029500.1 | - | 6 | 1150 | |||
AK077060.1 | - | 6 | 1111 |