Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018814.3 | NP_061284.2 | 28 | 5557 | ||
GenBank mRNA | AF096286.1 | - | 22 | 3387 | ||
AK014387.1 | - | 2 | 245 | |||
AK129221.1 | - | 23 | 3437 | |||
AK154122.1 | - | 9 | 1285 | |||
AK170320.1 | - | 9 | 1285 | |||
BC057333.1 | - | 21 | 3300 | |||
BC066849.1 | - | 23 | 3222 | |||
BC098370.1 | - | 23 | 3147 |