Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018811.6 | NP_061281.3 | 6 | 817 | ||
GenBank mRNA | AF201730.1 | - | 6 | 806 | ||
AF546701.1 | - | 6 | 818 | |||
AK008690.1 | - | 6 | 827 | |||
AK016296.1 | - | 4 | 478 | |||
AK139526.1 | - | 6 | 826 | |||
AK153642.1 | - | 6 | 864 | |||
BC027098.1 | - | 6 | 787 | 5356667 | ||
BC089477.1 | - | 6 | 797 | 6741521 |