Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf40b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018786.2 | NP_061256.1 | 18 | 1771 | ||
GenBank mRNA | AF135440.1 | - | 18 | 1771 | ||
AK140075.1 | - | 17 | 2084 | |||
AK164947.1 | - | 18 | 2128 | |||
BC051961.1 | - | 17 | 1750 | |||
BC058389.1 | - | 16 | 2675 | |||
BC082994.1 | - | 17 | 1929 | 6333535 |