Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npff | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018787.1 | NP_061257.1 | 2 | 145 | ||
GenBank mRNA | AF148701.1 | - | 2 | 145 | ||
BC043025.1 | - | 1 | 701 | |||
BC107294.1 | - | 1 | 375 | 40053067 | ||
BC107295.1 | - | 1 | 380 | 40053068 |