Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdgfc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019971.2 | NP_064355.1 | 5 | 1920 | ||
GenBank mRNA | AF117608.1 | - | 5 | 1096 | ||
AF266467.1 | - | 5 | 1920 | |||
AF286725.1 | - | 5 | 899 | |||
AK033734.1 | - | 5 | 1649 | |||
AK042767.1 | - | 5 | 1667 | |||
AK052947.1 | - | 5 | 1172 | |||
AK081347.1 | - | 6 | 972 | |||
BC029099.1 | - | 6 | 1223 | |||
BC037696.1 | - | 5 | 1107 | 4008749 |