Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gripap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290455.1 | NP_001277384.1 | 10 | 778 | ||
NM_207670.2 | NP_997553.1 | 10 | 778 | |||
GenBank mRNA | AK037997.1 | - | 4 | 1233 | ||
AK156165.1 | - | 10 | 779 | |||
AK158098.1 | - | 7 | 576 | |||
AK172590.1 | - | 8 | 644 | |||
AK172623.1 | - | 10 | 779 | |||
AK172699.1 | - | 10 | 779 | |||
AW342118.1 | - | 10 | 753 | |||
BC138523.1 | - | 10 | 774 | 8861543 | ||
BC145310.1 | - | 10 | 774 | 9053864 |