Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207202.2 | NP_997085.2 | 8 | 1003 | ||
GenBank mRNA | BC053918.1 | - | 8 | 968 | ||
BC139064.1 | - | 7 | 779 | 8862086 | ||
BU505792.1 | - | 3 | 213 |