Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081182.2 | NP_001074651.2 | 26 | 3253 | ||
GenBank mRNA | AF156546.1 | - | 4 | 512 | ||
AK155736.1 | - | 25 | 3287 | |||
AK173107.1 | - | 15 | 2698 |