Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173036.3 | NP_766624.3 | 9 | 1199 | ||
GenBank mRNA | AK037149.1 | - | 5 | 2144 | ||
AK154648.1 | - | 9 | 1175 | |||
AK155164.1 | - | 9 | 1171 | |||
BC026426.1 | - | 7 | 905 | |||
BC064726.1 | - | 9 | 1159 | 30433392 | ||
BC071188.1 | - | 7 | 1135 |
Set Membership: |
G-protein coupled receptor |