Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hes6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019479.3 | NP_062352.1 | 4 | 890 | ||
GenBank mRNA | AB035178.1 | - | 4 | 858 | ||
AF260236.1 | - | 4 | 862 | |||
AK005022.1 | - | 4 | 890 | |||
AK007480.1 | - | 4 | 898 | |||
AK012582.1 | - | 3 | 1129 | |||
AK013443.1 | - | 4 | 885 | |||
AK076135.1 | - | 4 | 890 | |||
AK146192.1 | - | 3 | 1095 | |||
AK155340.1 | - | 4 | 891 | |||
AK187013.1 | - | 1 | 126 | |||
AK217330.1 | - | 1 | 99 | |||
BC012897.1 | - | 4 | 868 | 4011223 |