Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018739.2 | NP_061209.1 | 5, 6 | 606 | ||
GenBank mRNA | AK146136.1 | - | 5, 6 | 587 | ||
AK168391.1 | - | 5, 6 | 599 | |||
BC048480.1 | - | 5, 6 | 464 | |||
BC096578.1 | - | 202 | ||||
BC115549.1 | - | 5, 6 | 566 | 40101700 | ||
BC115550.1 | - | 5, 6 | 566 | 40101701 | ||
D78255.1 | - | 5, 6 | 585 |