Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168568.1 | NP_001162040.1 | 15, 16 | 1670 | ||
NM_001168569.1 | NP_001162041.1 | 15, 16 | 1727 | |||
NM_018737.5 | NP_061207.1 | 15, 16 | 1802 | |||
GenBank mRNA | AK032018.1 | - | 15, 16 | 1529 | ||
AK052854.1 | - | 15, 16 | 1718 | |||
AK084595.1 | - | 15, 16 | 1599 | |||
AK085381.1 | - | 15, 16 | 1578 | |||
AK150966.1 | - | 15, 16 | 1669 | |||
AK151597.1 | - | 15, 16 | 1669 | |||
BC003257.1 | - | 15, 16 | 1510 | 3500778 | ||
EF410142.1 | - | 401 | ||||
U49385.1 | - | 15, 16 | 1692 |