Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018816.1 | NP_061286.1 | 4 | 390 | ||
GenBank mRNA | AF207820.1 | - | 4 | 390 | ||
AK004530.1 | - | 4 | 435 | |||
BC021597.1 | - | 4 | 427 | 5100655 |