Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap3m1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018829.4 | NP_061299.3 | 8, 9 | 1471 | ||
GenBank mRNA | AF242857.1 | - | 8, 9 | 1457 | ||
AK004740.1 | - | 8, 9 | 1217 | |||
BC010667.1 | - | 2, 3 | 354 | |||
BC024595.1 | - | 8, 9 | 1185 | 5026206 | ||
BC090983.1 | - | 8, 9 | 1219 | 30282230 |