Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclre1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018831.4 | NP_061301.3 | 2 | 824 | ||
GenBank mRNA | AF241240.1 | - | 1 | 304 | ||
AK012900.1 | - | 1 | 792 | |||
AK029794.1 | - | 1 | 1232 | |||
AK220214.1 | - | 1 | 52 | |||
BC072667.1 | - | 1 | 240 | 30629417 | ||
BC076617.1 | - | 2 | 824 |