Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclre1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018831.4 | NP_061301.3 | 2 | 951 | ||
GenBank mRNA | AF241240.1 | - | 1 | 431 | ||
AK012900.1 | - | 1 | 919 | |||
AK029794.1 | - | 1 | 1359 | |||
AK220214.1 | - | 1 | 179 | |||
BC072667.1 | - | 1 | 367 | 30629417 | ||
BC076617.1 | - | 2 | 951 |