Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclre1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018831.4 | NP_061301.3 | 7 | 3248 | ||
GenBank mRNA | AF241240.1 | - | 6 | 2713 | ||
AK029794.1 | - | 6 | 2917 | |||
AK220214.1 | - | 6 | 2470 | |||
BC072667.1 | - | 5 | 3846 | 30629417 | ||
BC076617.1 | - | 7 | 3248 |