Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000422.2 | NP_000413.1 | 1 | 527 | ||
GenBank mRNA | AK095342.1 | - | 1 | 456 | ||
AK304373.1 | - | 1 | 474 | |||
BC000159.2 | - | 1 | 456 | 2986100 | ||
BC011901.2 | - | 1 | 454 | 4111703 | ||
BC056421.1 | - | 1 | 456 | 5208304 | ||
BC072018.1 | - | 2 | 394 | 3531173 | ||
BC072019.1 | - | 1 | 452 | 5442690 | ||
BP226061.1 | - | 1 | 527 | |||
BX647923.1 | - | 1 | 193 | |||
X62571.1 | - | 1 | 454 |