Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018854.4 | NP_061342.1 | 5, 6 | 698 | ||
GenBank mRNA | AK002397.1 | - | 5, 6 | 699 | ||
AK139942.1 | - | 5, 6 | 735 | |||
AY082613.1 | - | 4, 5 | 418 | |||
BC002040.1 | - | 5 | 1051 | 3590790 | ||
U38981.1 | - | 531 |