Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpxm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018867.5 | NP_061355.3 | 6, 7 | 1045 | ||
GenBank mRNA | AF017639.1 | - | 6, 7 | 1046 | ||
AK019509.1 | - | 6, 7 | 1044 | |||
AK038874.1 | - | 6, 7 | 928 | |||
BC021444.1 | - | 6, 7 | 967 | 5038528 |