Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019488.4 | NP_062361.1 | 10 | 1422 | ||
GenBank mRNA | AF232061.2 | - | 10 | 1379 | ||
AJ245936.1 | - | 10 | 1399 | |||
AK081806.1 | - | 10 | 1439 | |||
AK141193.1 | - | 6 | 2462 | |||
AK155792.1 | - | 10 | 1452 | |||
AK170319.1 | - | 10 | 1435 | |||
AY856043.1 | - | 10 | 1383 | |||
AY856044.1 | - | 9 | 1274 | |||
AY856045.1 | - | 8 | 1103 | |||
AY856046.1 | - | 6 | 804 | |||
BC021758.1 | - | 6 | 748 | |||
BC090993.1 | - | 10 | 1422 | 6751763 | ||
Y17802.1 | - | 10 | 1404 |