Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019990.4 | NP_064374.1 | 7 | 938 | ||
GenBank mRNA | AB031550.1 | - | 8 | 1115 | ||
BC046335.1 | - | 7 | 1027 | |||
BC058773.1 | - | 7 | 1070 | |||
BC080808.1 | - | 7 | 1133 | |||
BC094007.1 | - | 7 | 951 | |||
BC116984.1 | - | 6 | 846 | 40126303 | ||
BC120641.1 | - | 6 | 846 | 40129564 | ||
BC144877.1 | - | 5 | 793 | |||
BY581159.1 | - | 1 | 152 |